ENA388700239 Enamine

4-(pyridin-3-yl)pyrrolidin-2-one

  • MDL number MFCD16653126
  • Cas Number : 55656-99-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 69...71
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121811

Safety Information

msds https://www.enaminestore.com/catalog/EN300-121811
Availability details for
Quantity
  • Availability
  • Add to Cart