ENA388700241 Enamine

4-methyl-4-phenylcyclohexan-1-one

  • MDL number MFCD10569281
  • Cas Number : 18932-33-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 41-43
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109872

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109872
Availability details for
Quantity
  • Availability
  • Add to Cart