ENA388700242 Enamine

2-methyl-2H,3H,3aH,4H,5H,6H-cyclopenta[c]pyrazol-3-one

  • MDL number MFCD14653671
  • Cas Number : 1043920-64-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 155-157
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123960

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123960
Availability details for
Quantity
  • Availability
  • Add to Cart