ENA388700256 Enamine

ethyl 2-sulfamoylpyridine-3-carboxylate

  • MDL number MFCD16093784
  • Cas Number : 247136-80-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 92...94
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123989

Safety Information

msds https://www.enaminestore.com/catalog/EN300-123989
Availability details for
Quantity
  • Availability
  • Add to Cart