ENA388700262 Enamine

2-methoxyquinolin-8-amine

  • MDL number MFCD19689979
  • Cas Number : 134829-04-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123995

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123995
Availability details for
Quantity
  • Availability
  • Add to Cart