ENA388700267 Enamine

2-[2-(4-methylphenoxy)acetamido]benzoic acid

  • MDL number MFCD00745997
  • Cas Number : 59090-64-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124001

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124001
Availability details for
Quantity
  • Availability
  • Add to Cart