ENA388700271 Enamine

5-(methoxycarbonyl)pyrazine-2-carboxylic acid

  • MDL number MFCD14581974
  • Cas Number : 1206250-26-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124007

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124007
Availability details for
Quantity
  • Availability
  • Add to Cart