ENA388700273 Enamine

(3-aminopropoxy)trimethylsilane

  • MDL number MFCD19203593
  • Cas Number : 23755-44-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124009

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124009
Availability details for
Quantity
  • Availability
  • Add to Cart