ENA388700276 Enamine

4-chloro-7-(diethylamino)-2-oxo-2H-chromene-3-carbaldehyde

  • MDL number MFCD00232978
  • Cas Number : 136918-66-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124036

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-124036
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart