ENA388700280 Enamine

2-[(4-methylphenyl)methyl]-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD14539664
  • Cas Number : 1216181-04-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124040

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124040
Availability details for
Quantity
  • Availability
  • Add to Cart