ENA388700282 Enamine

1,2,5-trimethylpiperazine

  • MDL number MFCD14584691
  • Cas Number : 82546-86-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124042

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124042
Availability details for
Quantity
  • Availability
  • Add to Cart