ENA388700286 Enamine

1-(trifluoromethyl)cyclohexan-1-ol

  • MDL number MFCD13659426
  • Cas Number : 80768-55-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124047

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124047
Availability details for
Quantity
  • Availability
  • Add to Cart