ENA388700289 Enamine

4-methyl-4-phenylcyclohex-2-en-1-one

  • MDL number MFCD00087385
  • Cas Number : 17429-36-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109873

Safety Information

msds https://www.enaminestore.com/catalog/EN300-109873
Availability details for
Quantity
  • Availability
  • Add to Cart