ENA388700293 Enamine

1-phenyl-3-(4-{[4-(trifluoromethyl)phenyl]methyl}piperazin-1-yl)propan-2-ol

  • MDL number MFCD23144163
  • Cas Number : 1427381-03-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124056

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124056
Availability details for
Quantity
  • Availability
  • Add to Cart