ENA388700294 Enamine

5-hydroxy-2-methyl-2,3-dihydro-1H-isoindol-1-one

  • MDL number MFCD12924811
  • Cas Number : 109803-65-8
  • Molecular Weight : 163.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 244-246
Iupac Name 5-hydroxy-2-methyl-1-isoindolinone
Inchi Code 1S/C9H9NO2/c1-10-5-6-4-7(11)2-3-8(6)9(10)12/h2-4,11H,5H2,1H3
Inchi Key IQMXSOJFRNHTRP-UHFFFAOYSA-N

Description

Melting Point 244-246
Purity 95
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-124057

Safety Information

pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart