ENA388700297 Enamine

2-amino-1-(pyridin-3-yl)ethan-1-one dihydrochloride

  • MDL number MFCD09260971
  • Cas Number : 51746-82-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124061

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124061
Availability details for
Quantity
  • Availability
  • Add to Cart