ENA388700298 Enamine

2-methyl-6-(6-methyl-1H-1,3-benzodiazol-2-yl)phenol

  • MDL number MFCD12069244
  • Cas Number : 1096325-53-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 244...246
Purity 95.00
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121973

Safety Information

msds https://www.enaminestore.com/catalog/EN300-121973
Availability details for
Quantity
  • Availability
  • Add to Cart