ENA388700299 Enamine

2-(2-tert-butyl-1,3-thiazol-4-yl)ethan-1-amine

  • MDL number MFCD17251056
  • Cas Number : 1266776-78-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122015

Safety Information

msds https://www.enaminestore.com/catalog/EN300-122015
Availability details for
Quantity
  • Availability
  • Add to Cart