ENA388700301 Enamine

methyl 2-[(2-amino-4-fluorophenyl)formamido]propanoate

  • MDL number MFCD18534025

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124079

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124079
Availability details for
Quantity
  • Availability
  • Add to Cart