ENA388700308 Enamine

8-methyl-1,8-diazaspiro[4.5]decane

  • MDL number MFCD12406535
  • Cas Number : 1153767-99-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124135

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124135
Availability details for
Quantity
  • Availability
  • Add to Cart