ENA388700312 Enamine

[2-(tert-butoxy)pyridin-4-yl]methanamine

  • MDL number MFCD10692481
  • Cas Number : 1152560-93-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124139

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124139
Availability details for
Quantity
  • Availability
  • Add to Cart