ENA388700315 Enamine

5-methyl-2,4-dihydro-1H-3,1-benzoxazine-2,4-dione

  • MDL number MFCD00797331
  • Cas Number : 20877-81-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 216-217
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124144

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124144
Availability details for
Quantity
  • Availability
  • Add to Cart