ENA388700319 Enamine

2-methyl-1-(1-methylpiperidin-4-yl)piperazine

  • MDL number MFCD19285682
  • Cas Number : 1267350-91-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124149

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124149
Availability details for
Quantity
  • Availability
  • Add to Cart