ENA388700321 Enamine

(2-chlorophenyl)(naphthalen-1-yl)methanone

  • MDL number MFCD00181818
  • Cas Number : 180780-15-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124151

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124151
Availability details for
Quantity
  • Availability
  • Add to Cart