ENA388700322 Enamine

3-(2,5-dimethyl-1H-pyrrol-1-yl)-4-methoxybenzoic acid

  • MDL number MFCD19182777
  • Cas Number : 1274929-77-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124153

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124153
Availability details for
Quantity
  • Availability
  • Add to Cart