ENA388700325 Enamine

tert-butyl 4-oxo-3,4-dihydrospiro[1-benzopyran-2,3'-pyrrolidine]-1'-carboxylate

  • MDL number MFCD15071774
  • Cas Number : 1013334-96-7
  • Molecular Weight : 303.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 4-oxospiro[chromane-2,3'-pyrrolidine]-1'-carboxylate
Inchi Code 1S/C17H21NO4/c1-16(2,3)22-15(20)18-9-8-17(11-18)10-13(19)12-6-4-5-7-14(12)21-17/h4-7H,8-11H2,1-3H3
Inchi Key VXPLDEWVQHLNAA-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-124159

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-124159
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart