ENA388700328 Enamine

4-(2-chlorophenyl)-1,2,3,6-tetrahydropyridine hydrochloride

  • MDL number MFCD23144171
  • Cas Number : 97429-95-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124163

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124163
Availability details for
Quantity
  • Availability
  • Add to Cart