ENA388700334 Enamine

4,5-dihydro-1H-pyrazol-3-amine hydrochloride

  • MDL number MFCD23144174
  • Cas Number : 82845-80-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-124177

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124177
Availability details for
Quantity
  • Availability
  • Add to Cart