ENA388700340 Enamine

4-amino-1-methyl-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD05667196
  • Cas Number : 790164-97-5
  • Molecular Weight : 141.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-124195

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124195
Availability details for
Quantity
  • Availability
  • Add to Cart