ENA388700342 Enamine

{1-[(4-fluorophenyl)methyl]cyclopropyl}methanol

  • MDL number MFCD21671025
  • Cas Number : 1427381-05-2
  • Molecular Weight : 180.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [1-(4-fluorobenzyl)cyclopropyl]methanol
Inchi Code 1S/C11H13FO/c12-10-3-1-9(2-4-10)7-11(8-13)5-6-11/h1-4,13H,5-8H2
Inchi Key ZMYSHCXWFHJOIL-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-124197

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-124197
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart