ENA388700354 Enamine

5-chloro-3-fluoro-2-hydroxybenzaldehyde

  • MDL number MFCD16658025
  • Cas Number : 394-96-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124214

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124214
Availability details for
Quantity
  • Availability
  • Add to Cart