ENA388700361 Enamine

4-[(2-methylphenyl)methoxy]piperidine

  • MDL number MFCD08688174
  • Cas Number : 81151-51-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124222

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124222
Availability details for
Quantity
  • Availability
  • Add to Cart