ENA388700365 Enamine

4-amino-1-methoxybutan-2-ol

  • MDL number MFCD19614231
  • Cas Number : 39754-69-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122346

Safety Information

msds https://www.enaminestore.com/catalog/EN300-122346
Availability details for
Quantity
  • Availability
  • Add to Cart