ENA388700377 Enamine

6-amino-1-(cyclohexylmethyl)-1,2,3,4-tetrahydropyrimidine-2,4-dione

  • MDL number MFCD12166806
  • Cas Number : 75914-78-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124263

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124263
Availability details for
Quantity
  • Availability
  • Add to Cart