ENA388700379 Enamine

{4-[4-(cyclopropylmethyl)piperazin-1-yl]phenyl}methanamine

  • MDL number MFCD10033297
  • Cas Number : 1018613-28-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124265

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124265
Availability details for
Quantity
  • Availability
  • Add to Cart