ENA388700386 Enamine

3,4-diethylaniline

  • MDL number MFCD18971287
  • Cas Number : 54675-14-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124282

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124282
Availability details for
Quantity
  • Availability
  • Add to Cart