ENA388700388 Enamine

2-phenyl-2H-1,2,3-triazol-4-amine

  • MDL number MFCD04000417
  • Cas Number : 39102-26-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124287

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124287
Availability details for
Quantity
  • Availability
  • Add to Cart