ENA388700393 Enamine

1-phenyl-1H-1,2,3-triazol-4-amine

  • MDL number MFCD20691554
  • Cas Number : 2076-64-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 103-105
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124295

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124295
Availability details for
Quantity
  • Availability
  • Add to Cart