ENA388700397 Enamine

4,5-dibromopyridin-3-amine

  • MDL number MFCD23144187
  • Cas Number : 1427379-89-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124300

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124300
Availability details for
Quantity
  • Availability
  • Add to Cart