ENA388700399 Enamine

N-(propan-2-yl)-5,6,7,8-tetrahydroquinolin-8-amine

  • MDL number MFCD21790639
  • Cas Number : 1427380-60-6
  • Molecular Weight : 190.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-isopropyl-5,6,7,8-tetrahydro-8-quinolinamine
Inchi Code 1S/C12H18N2/c1-9(2)14-11-7-3-5-10-6-4-8-13-12(10)11/h4,6,8-9,11,14H,3,5,7H2,1-2H3
Inchi Key MMIUIJWTEXAJOA-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-124303

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124303
Availability details for
Quantity
  • Availability
  • Add to Cart