ENA388700407 Enamine

ethyl 6-[(4-aminocyclohexyl)oxy]pyridine-3-carboxylate

  • MDL number MFCD17388919
  • Cas Number : 1274530-16-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-124331

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124331
Availability details for
Quantity
  • Availability
  • Add to Cart