ENA388700408 Enamine

2-(4-chloropyridin-2-yl)acetonitrile

  • MDL number MFCD09924172
  • Cas Number : 1000515-40-1
  • Molecular Weight : 152.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100629

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-100629
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart