ENA388700411 Enamine

2-[2-(trifluoromethyl)phenyl]cyclopentan-1-amine

  • MDL number MFCD17223380
  • Cas Number : 1343819-12-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124336

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124336
Availability details for
Quantity
  • Availability
  • Add to Cart