ENA388700413 Enamine

ethyl 1-isocyanatocyclobutane-1-carboxylate

  • MDL number MFCD23144192
  • Cas Number : 106366-62-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124338

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124338
Availability details for
Quantity
  • Availability
  • Add to Cart