ENA388700414 Enamine

1-(3,4-difluorophenyl)-2-methylpropan-1-ol

  • MDL number MFCD12153595
  • Cas Number : 1184394-61-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124341

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124341
Availability details for
Quantity
  • Availability
  • Add to Cart