ENA388700424 Enamine

3-tert-butyl-1-methyl-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD11180144
  • Cas Number : 1152509-69-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-124366

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124366
Availability details for
Quantity
  • Availability
  • Add to Cart