ENA388700425 Enamine

3-tert-butyl-1-methyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD11103105
  • Cas Number : 1152548-57-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140...142
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124367

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124367
Availability details for
Quantity
  • Availability
  • Add to Cart