ENA388700427 Enamine

1-methyl-3-(thiophen-2-yl)-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD11103099
  • Cas Number : 689251-87-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 64...66
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124369

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124369
Availability details for
Quantity
  • Availability
  • Add to Cart