ENA388700430 Enamine

2-(2,2,2-trifluoroethyl)-1,2,3,4-tetrahydroisoquinolin-5-amine

  • MDL number MFCD12778186
  • Cas Number : 1183387-63-4
  • Molecular Weight : 230.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124373

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124373
Availability details for
Quantity
  • Availability
  • Add to Cart