ENA388700432 Enamine

4-(2,5-dihydro-1H-pyrrol-3-yl)-N,N-dimethylaniline

  • MDL number MFCD20689059
  • Cas Number : 791581-28-7
  • Molecular Weight : 188.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-[4-(2,5-dihydro-1H-pyrrol-3-yl)phenyl]-N,N-dimethylamine
Inchi Code 1S/C12H16N2/c1-14(2)12-5-3-10(4-6-12)11-7-8-13-9-11/h3-7,13H,8-9H2,1-2H3
Inchi Key BAFAQAMERJAIND-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-124375

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124375
Availability details for
Quantity
  • Availability
  • Add to Cart